High content cell imaging system, ImageXpress Micro 4
Supplier: Molecular Devices
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5039859 MICRO4 5039856 5039857 5039858 5037556
MLDV5039859EA 2042 GBP
MLDV5039859 MLDVMICRO4 MLDV5039856 MLDV5039857 MLDV5039858 MLDV5037556
High content cell imaging system, ImageXpress Micro 4
Microscopes
The ImageXpress Micro 4 high content imaging system represents the culmination of four generations of imaging expertise. The agile design allows you to boost your research with this faster than ever system, while providing the option to upgrade to confocal in the future to align with your research needs.
  • Tackle a wide range of assays with oil and air objectives ranging from 1× to 100×
  • Automated five-position filter cube changer to facilitate multiplex assays with up to five filter sets and four objectives for a single experiment
  • Optically-encoded linear
More Product Information