Microtube racks, reversible
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8660 8670 8680 8601
700-2824EA 0 GBP
700-2824 700-2825 700-2826 MLBP8601
Microtube racks, reversible
Racks Tube Racks
Made of PP.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Sturdy
These racks are for microcentrifuge tubes with molded alphanumeric grid and also assorted colours are available.