Water purification system, Elix® Essential
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ZLXE0100BE ZLXE0050DK ZLXE0030FR ZLXE0030GE ZLXE0030IT ZLXE0030SP ZLXE0030SZ ZLXE0030DK ZLXE0050BE ZLXE0030BE ZLXE0050EU ZLXE0050FR ZLXE0050GE ZLXE0050IT ZLXE0050SP ZLXE0050SZ ZLXE0030UK ZLXE0030EU ZLXE0100DK ZLXE0050UK ZLXEV050DK ZLXEV100BE ZLXEV030FR ZLXEV030GE ZLXEV030IT ZLXEV030SP ZLXEV030SZ ZLXEV030DK ZLXEV050BE ZLXEV050EU ZLXEV050FR ZLXEV050GE ZLXEV050IT ZLXEV050SP ZLXEV050SZ ZLXEV030UK ZLXE0150DK ZLXE0100EU ZLXE0100FR ZLXE0100GE ZLXE0100IT ZLXE0100SP ZLXE0100SZ ZLXEV030EU ZLXE0150BE ZLXEV100DK ZLXE0150EU ZLXE0150FR ZLXE0150GE ZLXE0150IT ZLXE0150SP ZLXE0150SZ ZLXE0150UK ZLXE0100UK ZLXEV050UK ZLXEV150DK ZLXEV100EU ZLXEV100FR ZLXEV100GE ZLXEV100IT ZLXEV100SP ZLXEV100SZ ZLXEV150BE ZLXEV150EU ZLXEV150FR ZLXEV150GE ZLXEV150IT ZLXEV150SP ZLXEV150SZ ZLXEV150UK ZLXEV100UK
MILLZLXE0100BEEA 6100 GBP
MILLZLXE0100BE MILLZLXE0050DK MILLZLXE0030FR MILLZLXE0030GE MILLZLXE0030IT MILLZLXE0030SP MILLZLXE0030SZ MILLZLXE0030DK MILLZLXE0050BE MILLZLXE0030BE MILLZLXE0050EU MILLZLXE0050FR MILLZLXE0050GE MILLZLXE0050IT MILLZLXE0050SP MILLZLXE0050SZ MILLZLXE0030UK MILLZLXE0030EU MILLZLXE0100DK MILLZLXE0050UK MILLZLXEV050DK MILLZLXEV100BE MILLZLXEV030FR MILLZLXEV030GE MILLZLXEV030IT MILLZLXEV030SP MILLZLXEV030SZ MILLZLXEV030DK MILLZLXEV050BE MILLZLXEV050EU MILLZLXEV050FR MILLZLXEV050GE MILLZLXEV050IT MILLZLXEV050SP MILLZLXEV050SZ MILLZLXEV030UK MILLZLXE0150DK MILLZLXE0100EU MILLZLXE0100FR MILLZLXE0100GE MILLZLXE0100IT MILLZLXE0100SP MILLZLXE0100SZ MILLZLXEV030EU MILLZLXE0150BE MILLZLXEV100DK MILLZLXE0150EU MILLZLXE0150FR MILLZLXE0150GE MILLZLXE0150IT MILLZLXE0150SP MILLZLXE0150SZ MILLZLXE0150UK MILLZLXE0100UK MILLZLXEV050UK MILLZLXEV150DK MILLZLXEV100EU MILLZLXEV100FR MILLZLXEV100GE MILLZLXEV100IT MILLZLXEV100SP MILLZLXEV100SZ MILLZLXEV150BE MILLZLXEV150EU MILLZLXEV150FR MILLZLXEV150GE MILLZLXEV150IT MILLZLXEV150SP MILLZLXEV150SZ MILLZLXEV150UK MILLZLXEV100UK
Water purification system, Elix® Essential
Water Purification Systems Pure and Ultra Pure Water Systems
Elix® Essential delivers Type 2 water customised to fit a broad range of research areas. It is designed to be fed from potable water and is the ideal feed for any laboratory equipment, including ultrapure water systems such as Milli-Q® systems.
  • Depending on daily volume needs, different production flow rates are available (3, 5, 10 & 15 l/h)
  • Pure water is stored in a polyethylene tank designed to minimise risks of contamination during water storage
  • Vent filter provides effective protection against airborne contaminants; ASM (Automatic Sanitization
More Product Information