Epifluorescence Filter Holder
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
XF3001200 XF2014725
MILFXF3001200EA 1230 GBP
MILFXF3001200 MILFXF2014725
Epifluorescence Filter Holder
Filter Holders
Epifluorescence filter holder with stainless steel funnel and base, anodised aluminum spring clamp and silicone stopper.
  • Ideal for bacteriological analysis by epifluorescence microscopy
Designed for accurate, quantitative vacuum filtration of samples for bacteriological analysis. The highly polished stainless-steel funnel prevents bacteria cells from adhering to the surface.