MultiScreen® HTS Filter Plates, Styrene
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MSNANLY50 MSNANLY10
MILFMSNANLY50EA 1970 GBP
MILFMSNANLY50 MILFMSNANLY10
MultiScreen® HTS Filter Plates, Styrene
Microplates Filter Plates
MultiScreen plates have a long history as a reliable tool for the advancement of drug discovery and life science research.
  • Enhanced handling is achieved with rigid sidewalls
  • Abundant surface area for barcode labels
  • Prevents any cross-contamination during incubation
  • High throughput screening
  • Color code: Clear
MultiScreen HTS plates are specifically developed for high throughput use with automated workstations. Plasmid purification is a widely employed method in molecular cloning, transformation, and recombinant protein expression. Plasmid purificationMore Product Information