Undercounter laboratory glasswashers, PG 8583 CD
Supplier: MIELE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9960950 10224820 10224830 10224270 10224290 10224280 10203060 10224300
MIEL9960950EA 14200 GBP
MIEL9960950 MIEL10224820 MIEL10224830 MIEL10224270 MIEL10224290 MIEL10224280 MIEL10203060 MIEL10224300
Undercounter laboratory glasswashers, PG 8583 CD
Glassware Washing Machines
Miele PG 8583 CD Laboratory glasswasher represents a professional solution for laboratory glassware for analytical experiments. This approach is particularly gentle on materials and is recommended by leading manufacturers of laboratory glassware.
  • 'Touch on metal' controls with three-line text display such as program name, actual/target temperature and remaining program time
  • Program catering for all disciplines and vacant program slots for user-specific programs
  • Freely assignable direct access buttons and individually selectable program names for fast
More Product Information