Cylinder brushes
Supplier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BRWH-02 BRWH-03
MICRBRWH-02EA 104 GBP
MICRBRWH-02 MICRBRWH-03
Cylinder brushes
Brushes
These cylinder brushes with brush heads feature ACME thread for attachment to STA series handles.
  • With optional fabric covers
  • Polyester bristles
  • BPA-free
Base resins used to produce blocks and bristles are made from FDA approved materials.
Polyester cover is compatible with strong disinfectants and autoclavable.