May Grunwald's eosin-methylene blue solution, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ500 (more info.)

MG1L-1L
MG1L-1LEA 202 GBP
MG1L-1L
May Grunwald's eosin-methylene blue solution, Sigma-Aldrich®
May Grunwald's eosin-methylene blue
Storage Temperature: Ambient
MDL Number: MFCD00131580
UN: 1230
ADR: 3,II
IVDLogo

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR