Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Tygon® S3™ E-Food, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX96458-12EA 96.3 GBP
MFLX96458-12 MFLX96458-26 MFLX96458-42 MFLX96458-18 MFLX96458-30 MFLX96458-34 MFLX96458-48
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Tygon® S3™ E-Food, Avantor®
Tubing Peristaltic Pump Tubing
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Specially designed for use with food applications
  • Extremely smooth bore
To complete your Ismatec® pumping system, order three-stop color-coded tubing and extension tubing. Tubing comes inMore Product Information