Masterflex® L/S® 2-Stop Pump Tubing, Chem-Durance® Bio, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX96117-13EA 171 GBP
MFLX96117-13 MFLX96117-14 MFLX96117-15 MFLX96117-16 MFLX96117-25 MFLX96117-35
Masterflex® L/S® 2-Stop Pump Tubing, Chem-Durance® Bio, Avantor®
Tubing Peristaltic Pump Tubing
Excellent chemical resistance and long pumping life.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for
More Product Information