Masterflex® L/S® 2-Stop Pump Tubing, Biopharm Plus Platinum-Cured Silicone, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX96116-13EA 158 GBP
MFLX96116-13 MFLX96116-14 MFLX96116-15 MFLX96116-16 MFLX96116-24 MFLX96116-25 MFLX96116-35
Masterflex® L/S® 2-Stop Pump Tubing, Biopharm Plus Platinum-Cured Silicone, Avantor®
Tubing Peristaltic Pump Tubing
Provides longer life than other platinum and peroxide silicone tubing formulations.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include
More Product Information