Masterflex® L/S® Sanitary Fitting Pump Tubing Assembly, PharMed® BPT, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX96102-17EA 202 GBP
MFLX96102-17 MFLX96102-18 MFLX96102-24 MFLX96102-35 MFLX96102-36 MFLX96103-14 MFLX96103-15 MFLX96103-16 MFLX96103-17 MFLX96103-18 MFLX96103-24 MFLX96103-25 MFLX96103-36 MFLX96102-13 MFLX96102-14 MFLX96102-15 MFLX96102-16 MFLX96102-25 MFLX96103-13
Masterflex® L/S® Sanitary Fitting Pump Tubing Assembly, PharMed® BPT, Avantor®
Tubing Peristaltic Pump Tubing
1/2" mini-clamp sanitary fittings are molded onto each end of the assembly for easy implementation into your high-purity system.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique
More Product Information