Hubtron™ Black Pascal Fluoroelastomer Tubing
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
95257-02 95257-06
MFLX95257-04EA 1090 GBP
MFLX95257-04 MFLX95257-02 MFLX95257-06
Hubtron™ Black Pascal Fluoroelastomer Tubing
Tubing Transfer Tubing
Ideal for corrosive chemical transfer.
  • Global market leader in range of formulations for most extensive chemical compatibility
Applications: Extremely demanding corrosive chemical transfer, electronics applications, ink and printing, and general manufacturing applications requiring UV protection.
Characteristics: Opaque black with UV block, extremely low gas permeability, extremely strong chemical resistance. Superior pressure ratings. Excellent chemical resistance to solvents, acids, bases, alcohols, and esters.
Certification: None.
Temperature: 14 to 30More Product Information