Masterflex® Ismatec® Reglo Independent Channel Control (ICC) Peristaltic Pumps, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX78001-58-EUEA 3110 GBP
MFLX78001-58-EU MFLX78001-60-EU MFLX78001-62-EU MFLX78001-68-EU MFLX78001-70-EU MFLX78001-78-EU MFLX78001-80-EU MFLX78001-82-EU MFLX78001-58-UK MFLX78001-60-UK MFLX78001-62-UK MFLX78001-68-UK MFLX78001-70-UK MFLX78001-72-UK MFLX78001-78-UK MFLX78001-80-UK MFLX78001-82-UK
Masterflex® Ismatec® Reglo Independent Channel Control (ICC) Peristaltic Pumps, Avantor®
Pumps Peristaltic Pumps
Independently control flow and direction of each channel via keypad or your PC.
  • Flow rate 0.001 to 43 ml/min per channel - flow rate depends on drive rpm, rollers, and tubing size
  • USB interface for quick connection to a PC
  • Flexibility of bidirectional flow in each channel
These multi-channel peristaltic pumps let you individually control the flow and direction of each channel from the intuitive keypad or via your computer—expanding your benchtop pumping capabilities with a single compact unit! Pump delivers continuous pumping or precision dispensingMore Product Information