Masterflex® L/S® Easy-Load® II Long-Shaft Pump Heads, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX77200-80EA 629 GBP
MFLX77200-80 MFLX77200-82
Masterflex® L/S® Easy-Load® II Long-Shaft Pump Heads, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Automatic retention holds tubing securely in place with no manual adjustment—just load the tubing and start the pump.
  • New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
  • Four-roller rotor improves pressure performance and mechanical stability while reducing pulsation
  • Model available with integrated open-head sensor–connects directly to sensor-equipped drives
  • Pump heads are stackable for multichannel applications
  • Compact design for efficient use of space
This L/S® Easy-Load® II PumpMore Product Information