Suction Line Pump Filter Strainers
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
74604-02 74604-04 74604-10 74604-12 74604-14 74604-20 74604-22 74604-23 74604-24 74604-30 74604-32 74604-34 74604-42 74604-43 74604-44 74604-52 74604-53 74604-54 74605-12
MFLX74604-02EA 11.4 GBP
MFLX74604-02 MFLX74604-04 MFLX74604-10 MFLX74604-12 MFLX74604-14 MFLX74604-20 MFLX74604-22 MFLX74604-23 MFLX74604-24 MFLX74604-30 MFLX74604-32 MFLX74604-34 MFLX74604-42 MFLX74604-43 MFLX74604-44 MFLX74604-52 MFLX74604-53 MFLX74604-54 MFLX74605-12
Suction Line Pump Filter Strainers
Filters Strainers
Protect pump from sediments filtering into suction line.
  • PVC (polyvinyl chloride) or Nylon base material of construction
  • 20, 40, 80 mesh sizes available
  • Easily removable for cleaning or replacing
These suction line pump filter strainers are ideal for keeping sediment and debris out of your pump for optimal performance. Choose from PVC (polyvinyl chloride) or nylon base material in a wide range of mesh sizes for any application.