Micropump® A-Mount Suction Shoe Gear Pump Heads
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
07003-02
MFLX07002-14EA 1490 GBP
MFLX07002-14 MFLX07002-25 MFLX07002-26 MFLX07002-27 MFLX07003-04 MFLX73003-14 MFLX73004-00 MFLX73004-02 MFLX73004-04 MFLX74004-19 MFLX74004-21 MFLX74004-77 MFLX07003-02 MFLX74012-51
Micropump® A-Mount Suction Shoe Gear Pump Heads
Pumps Pump Heads Gear Pump Heads
Smooth pulseless flow provides precise, leak-free delivery of fluids.
  • Suction shoe design provides higher delivery pressures and temperatures, plus increased efficiency.
Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and static seals keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of materials (due to wetted parts). Pump heads are compact and easy to service. Modular design offers flexibility—configure to your specific needs.