Micropump® A-Mount Cavity-Style Pump Heads
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX07001-40EA 1320 GBP
MFLX07001-40 MFLX07001-80 MFLX07002-17 MFLX07002-23 MFLX73011-00 MFLX73011-02 MFLX73011-08 MFLX73011-16 MFLX73011-18 MFLX74012-21
Micropump® A-Mount Cavity-Style Pump Heads
Pumps Pump Heads Gear Pump Heads
Smooth pulseless flow provides precise, leak-free delivery of fluids.
  • Cavity-style pump heads are designed to handle higher inlet pressures.
Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and static seals keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of materials (due to wetted parts). Pump heads are compact and easy to service. Modular design offers flexibility—configure to your specific needs.