Polypropylene (PP) Moderate-Flow Chemical-Resistance Centrifugal Pumps
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
72010-00 72010-05 72010-10 72010-15 72010-20 72010-25 72010-30 72010-35 72010-40 72010-45 72010-50 72010-60 72010-65 72010-80 72010-70
MFLX72010-00EA 0 GBP
MFLX72010-00 MFLX72010-05 MFLX72010-10 MFLX72010-15 MFLX72010-20 MFLX72010-25 MFLX72010-30 MFLX72010-35 MFLX72010-40 MFLX72010-45 MFLX72010-50 MFLX72010-60 MFLX72010-65 MFLX72010-80 MFLX72010-70
Polypropylene (PP) Moderate-Flow Chemical-Resistance Centrifugal Pumps
Pumps Centrifugal Pumps
Twin bearing system handles thrust loads for extended service life.
  • Quiet, energy efficient, and compact
  • Internal cooling circuit reduces bearing temperature
  • Enclosed motor for use in harsh environments
  • All wetted parts are nonmetallic for excellent chemical resistance
Moderate-Flow Polypropylene Pumps are an ideal option for process, recirculation, or transfer applications where continuous flow and excellent chemical resistance are needed.
Delivery information: Includes a 6-ft (1.8 m) cord with stripped ends.