Masterflex® Mechanically Coupled High-Head Centrifugal Pumps, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
70725-13 70725-11 70725-20 70725-22 70725-26 70725-31 70725-32 70725-36 70725-37
MFLX70725-13EA 1130 GBP
MFLX70725-13 MFLX70725-11 MFLX70725-20 MFLX70725-22 MFLX70725-26 MFLX70725-31 MFLX70725-32 MFLX70725-36 MFLX70725-37
Masterflex® Mechanically Coupled High-Head Centrifugal Pumps, Avantor®
Pumps Centrifugal Pumps
Rugged continuous-duty service with chemical and corrosion resistance.
  • Close-coupled design saves space; simplifies maintenance and installation
  • Stainless steel liquid end components deliver corrosion resistance
  • Versatile mounting can be installed horizontally or vertically
  • Back-pullout construction assembly allows overhaul of impeller and seal without disturbing suction and discharge connections
  • Top centerline discharge and foot support under casing ensures self-venting and reduces misalignment from pipe loads
  • High operating efficiency lowers operating
More Product Information