Masterflex® Complete Drum and Container Pumps, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
70613-00 70613-10 70613-20 70613-30 70613-40 70613-45 70613-50 70613-05 70613-25 70613-35 70613-55
MFLX70613-00EA 0 GBP
MFLX70613-00 MFLX70613-10 MFLX70613-20 MFLX70613-30 MFLX70613-40 MFLX70613-45 MFLX70613-50 MFLX70613-05 MFLX70613-25 MFLX70613-35 MFLX70613-55
Masterflex® Complete Drum and Container Pumps, Avantor®
Pumps Drum/Transfer Pumps
System includes pump tube and motor—tube material options ensure chemical compatibility.
  • Flow rate 35 GMP for all models
  • Equipped with variable-speed control motors (except air and explosion-proof models) for added safety and control
  • Maintenance is reduced with no mechanical seals
  • Run dry capability
Drum and container pumps are engineered to safely transfer a variety of liquids from drums, tanks, and small storage vessels. Materials such as PP, PVDF, CPVC, and 316 SS provide excellent chemical resistance against corrosive liquids. Order optionalMore Product Information