Masterflex® Adapter Fittings, Female Luer to Hose Barb, Bulkhead, Polypropylene, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX45508-30EA 30.6 GBP
MFLX45508-30 MFLX45508-32 MFLX45508-34
Masterflex® Adapter Fittings, Female Luer to Hose Barb, Bulkhead, Polypropylene, Avantor®
Fittings and Connectors
Ensure your process integrity with full material traceability.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options
More Product Information