Masterflex® 3/4" Sanitary Clamp Medium Flow Female Quick-Disconnect Fittings, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX40257-00EA 144 GBP
MFLX40257-00 MFLX40257-01 MFLX40257-05 MFLX40257-06 MFLX40257-10 MFLX40257-11 MFLX40257-15 MFLX40257-16 MFLX40257-20 MFLX40257-21 MFLX40257-25 MFLX40257-26 MFLX40257-30 MFLX40257-31 MFLX40257-35 MFLX40257-36
Masterflex® 3/4" Sanitary Clamp Medium Flow Female Quick-Disconnect Fittings, Avantor®
Fittings and Connectors
Mechanical innovations reduce risk of fluid cross-over and contamination. A mating connection is made with a Medium Flow Male fitting.
  • Aseptic connect and disconnect
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to pass through at lower line pressures in a smaller body. As the overall quick disconnect design has matured,More Product Information