Masterflex® 3/4" Sanitary Clamp High Flow Female Quick-Disconnect Fittings, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX40257-02EA 153 GBP
MFLX40257-02 MFLX40257-03 MFLX40257-04 MFLX40257-07 MFLX40257-08 MFLX40257-09 MFLX40257-12 MFLX40257-13 MFLX40257-14 MFLX40257-17 MFLX40257-18 MFLX40257-19 MFLX40257-22 MFLX40257-23 MFLX40257-24 MFLX40257-27 MFLX40257-28 MFLX40257-29 MFLX40257-32 MFLX40257-33 MFLX40257-34 MFLX40257-37 MFLX40257-38 MFLX40257-39
Masterflex® 3/4" Sanitary Clamp High Flow Female Quick-Disconnect Fittings, Avantor®
Fittings and Connectors
Mechanical innovations reduce risk of fluid cross-over and contamination. A mating connection is made with a High Flow Male fitting.
  • Aseptic connect and disconnect
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to pass through at lower line pressures in a smaller body. As the overall quick disconnect design has matured, aMore Product Information