Masterflex® Flowmeters and Controllers for Water, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
32907-43 32907-44 32908-45 32908-47 32907-45 32907-46 32908-41 32908-42 32908-43 32908-44 32908-46 32908-48 32908-49
MFLX32907-43EA 2680 GBP
MFLX32907-43 MFLX32907-44 MFLX32908-45 MFLX32908-47 MFLX32907-45 MFLX32907-46 MFLX32908-41 MFLX32908-42 MFLX32908-43 MFLX32908-44 MFLX32908-46 MFLX32908-48 MFLX32908-49
Masterflex® Flowmeters and Controllers for Water, Avantor®
Flow Switches
Achieve accurate, ultrafast volumetric measurement and control.
  • Technology allows for installation without typical inlet/outlet straight-run requirements
  • Units provide data on flow and temperature
  • All functions are user-programmable through an integrated keypad
These meters measure flow via pressure drop across a laminar flow element (LFE). Because the flow element makes the flow stream laminar, placement in the process does not require straight pipe runs upstream or downstream of the meter, greatly simplifying installation. The LFEs also provide anMore Product Information