Masterflex® Sanitary Clamps, Nylon, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX31201-93EA 28.1 GBP
MFLX31201-93 MFLX31201-95 MFLX31201-97 MFLX31201-99
Masterflex® Sanitary Clamps, Nylon, Avantor®
Clamps Tubing and Sanitary Clamps
Compatible with metal and plastic Tri-Clamp® fittings.
  • Excellent chemical resistance
  • Rated to 150 psi
  • Sterilizable via Autoclave and Gamma Irradiation
  • Features anti-tamper zip-tie lock out
  • Manufactured and packaged in cleanroom
Constructed of nylon, these Tri-Clamp® sanitary clamps are perfect for single-use transfer lines in biopharma markets, plus a wide range of fluid transfer systems. These clamps ideal for securing your fluid transfer system without damaging the process lines.