Barblock Fitting and Tubing Tools and Accessories
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX30604-12EA 266 GBP
MFLX30604-12 MFLX30604-14 MFLX30604-16 MFLX30604-18
Barblock Fitting and Tubing Tools and Accessories
Puller and Inserter Tools
Easily install your Barblock Fittings.
  • Optimize performance of your Barblock fittings
Select a hand tool to assemble your BarbLock Retainers based on the tubing size. Universal removal tool is also available.