Masterflex® Gold-coloured Identifier Sanitary Clamps, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX30562-51EA 34 GBP
MFLX30562-51 MFLX30562-52 MFLX30562-53 MFLX30562-54 MFLX30562-55
Masterflex® Gold-coloured Identifier Sanitary Clamps, Avantor®
Clamps Tubing and Sanitary Clamps
Easy visual identification of critical joints for review in your process.
  • Constructed of gold-coloured 304 Stainless Steel
Easily distinguishable from standard stainless steel clamps from a distance, these gold-coloured sanitary clamps are identical in size and function and can be used interchangeably with standard stainless steel clamps. Clamps feature a TiN (titanium nitride) coating. Clamps are available in a variety of sizes.