Masterflex® Platinum-cured Silicone Screen Gaskets, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
30523-20 30523-24 30523-28 30523-40 30523-44 30523-60 30523-64
MFLX00253-BMEA 79.7 GBP
MFLX00253-BM MFLX00269-AW MFLX00431-JA MFLX30523-04 MFLX30523-06 MFLX30523-08 MFLX30523-22 MFLX30523-26 MFLX30523-42 MFLX30523-46 MFLX30523-48 MFLX30523-62 MFLX30523-66 MFLX30523-68 MFLX30523-80 MFLX30523-82 MFLX30523-84 MFLX30523-86 MFLX30523-88 MFLX30523-20 MFLX30523-24 MFLX30523-28 MFLX30523-40 MFLX30523-44 MFLX30523-60 MFLX30523-64 MFLX41000-50 MFLX00383-PO
Masterflex® Platinum-cured Silicone Screen Gaskets, Avantor®
Seals Gaskets
Protect your pump, valves and other components in fluid line.
  • Pre-filter before critical membrane filters
  • Generate turbulence for enhanced mass or heat transfer
  • Purify in chromatography applications
Screen gaskets are designed to be interchangeable with standard sanitary clamp gaskets. Gaskets are available in platinum-cured silicone, Viton®, EPDM, PTFE, and Tuf-Steel in mesh sizes of 10, 20, 40, 60 and 100. Screen mesh is constructed of 316L SS. Gaskets meet USP Class VI, USDA, 3-A and cGMP criteria. FDA compliant. All gaskets are animal-derivedMore Product Information