Masterflex® Strainer Bottoms for Low-Cost In-Line Strainer Systems, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
29595-61 29595-63 29595-67 29595-71 29595-73 29595-79
MFLX29595-61EA 20.6 GBP
MFLX29595-61 MFLX29595-63 MFLX29595-67 MFLX29595-71 MFLX29595-73 MFLX29595-79
Masterflex® Strainer Bottoms for Low-Cost In-Line Strainer Systems, Avantor®
Filters Strainers
Easily customize strainer system to suit your application.
  • Choose from either black polypropylene or clear nylon strainer bottoms
Build your own economical filtration system to filter out particulates from your fluids. Available in three classes: low-profile, intermediate, and large. Modify or change components of the same class to meet the demands of your application.
Low-Profile Strainers are ideal for small-scale low-volume filtration. Use in research applications, cleaning equipment, and laboratory equipment.
Intermediate Strainers feature a largeMore Product Information