Masterflex® DB-9 Pump Interface Cables, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX22050-54EA 27 GBP
MFLX22050-54
Masterflex® DB-9 Pump Interface Cables, Avantor®
Cables
Optimize performance of your Masterflex® pump.
  • Suitable for use with Masterflex® L/S® drives with a 9-pin connector
Use these 9-pin interface cable assemblies with Masterflex L/S drives having a 9-pin connector; including the 07550- and 07551-series computer-compatible drives. Cable measures 6 ft (1.8 m) long.