Masterflex® Nylon Clamps for Sanitary Fittings, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX00424-TNEA 264 GBP
MFLX00424-TN MFLX00424-TO MFLX12805-05
Masterflex® Nylon Clamps for Sanitary Fittings, Avantor®
Clamps Tubing and Sanitary Clamps
Collared clamp helps prevent unintentional opening.
  • All fittings are compatible with Tri-Clamp® fittings
  • Excellent chemical resistance; low leachable characteristics