Masterflex® L/S® Economy Fixed-Speed Drives, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX07540-01EA 522 GBP
MFLX07540-01 MFLX07540-02 MFLX07540-06 MFLX07540-12 MFLX07540-20 MFLX07540-30 MFLX07540-60 MFLX07542-01-EU MFLX07542-02-EU MFLX07542-06-EU MFLX07542-12-EU MFLX07542-20-EU MFLX07542-30-EU MFLX07542-60-EU MFLX07542-01-UK MFLX07542-02-UK MFLX07542-06-UK MFLX07542-12-UK MFLX07542-20-UK MFLX07542-30-UK MFLX07542-60-UK
Masterflex® L/S® Economy Fixed-Speed Drives, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Economical drives for reliable general transfer and feed applications.
  • Economy fixed-speed drives are stackable and ideal for lab, field, or process feed applications
  • Choose from seven speeds based on flow rate or number of heads required
  • Constructed with painted steel housing for IP22-rating
The Masterflex® L/S® economy fixed-speed drives are compact and ideal where space is limited. These drives feature a lighted power switch on the front of the drive that lets you know when the pump is running. They accept five different pump head types: Standard,More Product Information