Masterflex® I/P® Standard Pump Heads, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX07019-20EA 657 GBP
MFLX07019-20 MFLX07019-21 MFLX07019-31 MFLX07019-32 MFLX07019-43 MFLX07019-53
Masterflex® I/P® Standard Pump Heads, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Best accuracy and repeatability of any I/P pump head—ideal for dispensing and metering.
  • Each model is optimized for a single tube size to maximize performance and tubing life
  • Cost effective pump head is an excellent choice for dedicated applications
  • Stack two pump heads to double flow rate
The Standard pump head’s classic design ensures the most accurate and repeatable performance—important in dispensing applications. A 360-degree precision molded occlusion body ensures optimum accuracy and long tubing life.
Select from PC housing with CRS or SS rotor,More Product Information