Adapter Fittings, Female Threaded, Tee
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
06546-17 06546-18 06546-19 06546-20 34030-19 34030-20 34030-21 34030-22
MFLX06546-17EA 67.6 GBP
MFLX06546-17 MFLX06546-18 MFLX06546-19 MFLX06546-20 MFLX34030-19 MFLX34030-20 MFLX34030-21 MFLX34030-22
Adapter Fittings, Female Threaded, Tee
Fittings and Connectors
Maintain a permanent connection.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery