Metal Body Hand-Operated Lever Drum Pumps
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
06511-00 06511-05 06511-10
MFLX06511-00EA 108 GBP
MFLX06511-00 MFLX06511-05 MFLX06511-10
Metal Body Hand-Operated Lever Drum Pumps
Pumps Drum/Transfer Pumps
Metal-body pumps for use in tight work areas where drums containing flammables are stored.
  • Curved discharge spout for convenient hose extension
  • Designed for use in industrial, agricultural, and automotive industries
  • Model 06511-00 is ideal for use with high-viscosity fluids
  • Materials vary from Stainless steel to steel bodies with PTFE or Delrin acetal construction allow for easy handling of the toughest fluids
Lever drum pump model 06511-05 is able to dispense kerosene, antifreeze, lightweight oils, non-water-based and non-corrosive detergents, waxes,More Product Information