Hand-Operated Rotary Drum Pumps
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
06510-00 06510-05 06510-10 06510-17 06510-19
MFLX06510-00EA 146 GBP
MFLX06510-00 MFLX06510-05 MFLX06510-10 MFLX06510-17 MFLX06510-19
Hand-Operated Rotary Drum Pumps
Pumps Drum/Transfer Pumps
Dispense a variety of liquids compatible with polypropylene, Ryton® PPS, cast iron, aluminum, stainless steel, or PTFE.
  • Lightweight and easy to use
  • Smooth, non-pulsating flow
  • Operate either in forward or reverse direction with a discharge hose
Polypropylene rotary pump (06510-00) is excellent for most organic acids, waxes, plating solutions, detergents and water soluble inorganic chemicals. Ryton PPS rotary pump (06510-05) is able to dispense aggressive chemicals such as esters, ketones and hydrocarbons. Ryton PPS pumps include a stainless steelMore Product Information