Masterflex® L/S® Precision Pump Tubing, PharMed® BPT, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX06508-14EA 174 GBP
MFLX06508-14 MFLX06508-13 MFLX06508-16 MFLX06508-17 MFLX06508-18 MFLX06508-25
Masterflex® L/S® Precision Pump Tubing, PharMed® BPT, Avantor®
Tubing Peristaltic Pump Tubing
Protect your light-sensitive samples.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly
More Product Information