Masterflex® L/S® Precision Pump Tubing, PharmaPure®, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX06435-13EA 167 GBP
MFLX06435-13 MFLX06435-14 MFLX06435-16 MFLX06435-17 MFLX06435-18 MFLX06435-25
Masterflex® L/S® Precision Pump Tubing, PharmaPure®, Avantor®
Tubing Peristaltic Pump Tubing
Ultra low spallation protects fluid purity – ideal for sensitive and costly fluids and for filtration applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
More Product Information