Masterflex® L/S® High-Performance Precision Pump Tubing, C-Flex® ULTRA, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX06434-15EA 124 GBP
MFLX06434-15 MFLX06434-35 MFLX06434-36 MFLX06434-24 MFLX06437-24
Masterflex® L/S® High-Performance Precision Pump Tubing, C-Flex® ULTRA, Avantor®
Tubing Peristaltic Pump Tubing
Longest life of any C-Flex® formulation.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-
More Product Information