C-Flex® Transfer Tubing, Animal-Derived Component Free (ADCF)
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
06401-72 06432-56
MFLX06432-50EA 106 GBP
MFLX06432-50 MFLX06432-51 MFLX06432-52 MFLX06432-53 MFLX06432-54 MFLX06432-55 MFLX06432-57 MFLX06432-58 MFLX06432-59 MFLX06432-60 MFLX06432-61 MFLX06432-62 MFLX61063-76 MFLX06401-72 MFLX06432-56
C-Flex® Transfer Tubing, Animal-Derived Component Free (ADCF)
Tubing Transfer Tubing
Meets the demands of various industries.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Characteristics: Exceptional tensile and tear strength, good biocompatibility, excellent chemical resistance, low gas permeability, and a smooth surface. Transparent.
Certification: Resin meets USP and Class VI requirements.More Product Information