Masterflex® Transfer Tubing, C-Flex®, Opaque White, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX06424-03EA 63.7 GBP
MFLX06424-03 MFLX06424-12 MFLX06424-19 MFLX06424-59 MFLX06424-60 MFLX06424-62 MFLX06424-63 MFLX06424-64 MFLX06424-65 MFLX06424-66 MFLX06424-69 MFLX06424-71 MFLX06424-72 MFLX06424-74 MFLX06424-75 MFLX06424-79 MFLX06424-81 MFLX06424-67 MFLX06424-76
Masterflex® Transfer Tubing, C-Flex®, Opaque White, Avantor®
Tubing Transfer Tubing
Meets the demands of various industries.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Applications: General-purpose lab uses requiring high performance and long life.
Characteristics: Exceptional tensile and tear strength, good biocompatibility, excellent chemical resistance, low gas permeability, and aMore Product Information