Stopcocks with Flow Indicator, PVC
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
06225-52 06225-54 06225-56 06225-50
MFLX06225-52EA 42.2 GBP
MFLX06225-52 MFLX06225-54 MFLX06225-56 MFLX06225-50
Stopcocks with Flow Indicator, PVC
Stopcocks
Easily control flow with flow indicator handle.
  • Constructed of durable PVC and EPDM
PVC bodies, EPDM seals and seats. Flow indicator on handle. Maximum pressure is 150 psi at the maximum temperature of 120 °F (49 °C); maximum vacuum is 29.9" Hg.