Thermo Scientific™ Nalgene™ Two- and Three-Way Stopcocks with Hose Barb Connections
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
06222-20 06222-40 06225-20 06225-40
MFLX06222-20EA 153 GBP
MFLX06222-20 MFLX06222-40 MFLX06225-20 MFLX06225-40
Thermo Scientific™ Nalgene™ Two- and Three-Way Stopcocks with Hose Barb Connections
Stopcocks
Easily adjust flow.
  • Tubulated outlets are arranged in a T shape and accepts 1/4 through 7/16" tubing
  • Polypropylene, with non-seizing, Teflon™ TFE plug
  • Autoclavable
Experience in-house fluid transfer system flexibility with the two- and three-way stopcock, arranged in a straight (two-way) or T-shape (three-way) to connect any two — or all three — arms of the stopcock with ease