George Fischer Type 546 Ball Valves 42387
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
01357-01 01357-03 01357-05 01357-07 01371-08 01371-09 01374-92
MFLX01357-01EA 105 GBP
MFLX01357-01 MFLX01357-03 MFLX01357-05 MFLX01357-07 MFLX01371-08 MFLX01371-09 MFLX01374-92
George Fischer Type 546 Ball Valves 42387
Valves Ball Valves
Optimize safety with this true union ball valve.
  • True union ball valve with added safety and functionality for a diverse range of applications
  • PTFE ball seat with high abrasion resistance
  • Built-in tool for easy adjustment of ball torque
  • Threaded mounting inserts for fixed-point attachment
Ball valves are designed primarily for on/off service. These valves contain a ball with a hole through it. A handle or electronic actuator rotates the ball 90 degrees, turning the flow on or off.