Tubing, FEP
Supplier: METROHM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
6.1805.010 6.1805.080 6.1805.100 6.1805.020 6.1805.180 6.1805.060 6.1805.120 6.1829.020
METR6.1805.010EA 177 GBP
METR6.1805.010 METR6.1805.080 METR6.1805.100 METR6.1805.020 METR6.1805.180 METR6.1805.060 METR6.1805.120 METR6.1829.020
Tubing, FEP
Tubing Transfer Tubing
FEP, translucent.
  • With light and kink protection
  • With thread connections at both ends
Material allows much longer lengths and tighter tolerances than PTFE tubing.