TableWorx Performance Work Tables, 304 Stainless Steel Work Surface
Supplier: Metro International
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TWM3060SI304S TWM3072SI304S TWM3096SI304S
METOTWM3060SI304SEA 0 GBP
METOTWM3060SI304S METOTWM3072SI304S METOTWM3096SI304S
TableWorx Performance Work Tables, 304 Stainless Steel Work Surface
Furniture Tables and Workbenches
TableWorx is a truly modular platform of performance work tables designed to allow you to customise your perfect solution.
All stainless series TableWorx performance work tables feature a 16-gauge, type 304 stainless steel work surface reinforced with 16-gauge hat channels. The sound deadened work surface features 90° turned down edges on all four sides. All sizes are available with an optional 51 mm stainless steel backsplash, which is formed into the work surface.
All stainless series TableWorx performance work tables feature type 304 stainless steel legs and leg mounts. A sturdy,More Product Information