Divider and tote boxes for shelving systems
Supplier: Metro International
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
DL93080NAT DS93080NAT MTB93030W MTB93060W DL93030NAT DS93030NAT MTB93080W
METODL93080NATEA 11.7 GBP
METODL93080NAT METODS93080NAT METOMTB93030W METOMTB93060W METODL93030NAT METODS93030NAT METOMTB93080W
Divider and tote boxes for shelving systems
Trays/Dishes/Pots Drawer Organisation Trays
The tote box provide safe and efficient storage of items in bulk, or in compartmentalised fashion with the use of optional dividers and covers.
PP base resin.
  • Ideal for storage and can be transported
  • Available in various colour options
  • Injection molded for longer life and more uniform wall thickness