EasyFrit ES1210 EasySampler Filter Kit
Supplier: Mettler - Toledo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
30811887 30811900
METB30811887EA 0 GBP
METB30811887 METB30811900
EasyFrit ES1210 EasySampler Filter Kit
Filters Liquid Process Filters
EasyFrit turns EasySampler into a selective solution phase sampling tool for data-rich collection throughout a controlled crystallisation.
  • HPLC ready samples
  • Eliminate user safety concerns
  • Avoid surface fouling
  • Transform and increase productivity
  • Never miss a reaction event
  • Representative and reproducible
  • Material: FEP, alloy C-276, C22 alloy
A unique self-flushing mechanism prevents surface fouling and allows unattended sampling over an elongated period under reaction conditions at scale.
Unattended, representative sampling: EasyFrit is aMore Product Information