Precision Balance, L-Platform, XPR
Supplier: Mettler - Toledo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
30316292 30316485 30317428
METB30316292EA 0 GBP
METB30316292 METB30316485 METB30317428
Precision Balance, L-Platform, XPR
Balances and Scales
Precision balance with exceptional results and smart quality assurance.
  • Safe and easy cleaning
  • Outstanding weighing performance
  • Audit-Proof results every time
  • Management of users and routine tests and consistent workflows
  • StatusLight™ uses color to provide instant information on the balance status
The MonoBloc™ weighing cell delivers fast, accurate and reliable results, ensuring high productivity even in challenging weighing conditions. The StatusLight™, LevelControl, and GWP Approved provide at-a-glance information to verify your results are validMore Product Information